• USA Home
  • Search Results

  • sulfapyridine2492914483211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sulfapyridine

5 Products
Synonym:   4-Amino-N-[2-pyridyl]benzene sulfonamide, N1-(Pyridin-2-yl)sulfanilamide
Image of Sulfapyridine
CAS Number:
Empirical Formula (Hill Notation):
 C11H11N3O2S
Molecular Weight:
 249.29
 Beilstein/REAXYS Number:
 222065
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Anal. Standard Form
  Physical Form
Show More