• USA Home
  • Search Results

  • sulfathiazolesodiumsalt2773014474111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sulfathiazole sodium salt

1 Products
Synonym:   4-Amino-N-(2-thiazolyl)benzenesulfonamide sodium salt
Image of Sulfathiazole sodium salt
CAS Number:
Empirical Formula (Hill Notation):
 C9H8N3NaO2S2
Molecular Weight:
 277.30
 Beilstein/REAXYS Number:
 3802297
 EC Number:
 MDL number:
 NACRES:
 NA.85
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
  Mode of Action
Show More