• USA Home
  • Search Results

  • sulforhodaminebacidchloride577116279629611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sulforhodamine B acid chloride

1 Products
Synonym:   Lissamine rhodamine B sulfonylchloride
Image of Sulforhodamine B acid chloride
CAS Number:
Empirical Formula (Hill Notation):
 C27H29ClN2O6S2
Molecular Weight:
 577.11
 Beilstein/REAXYS Number:
 7734927
 EC Number:
 MDL number:
 NACRES:
 NA.32
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More