• USA Home
  • Search Results

  • sulfur34s33971396597411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sulfur-34S

2 Products
Image of Sulfur-<SUP>34</SUP>S
CAS Number:
Linear Formula:
 34S
Molecular Weight:
 33.97
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Isotopic Purity
Show More