• USA Home
  • Search Results

  • sulfurdioxidesolution8208778299211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sulfur dioxide solution

1 Products
Synonym:   Sulfurous acid, Sulfurous anhydride
Image of Sulfur dioxide solution
CAS Number:
Linear Formula:
 H2SO3
Molecular Weight:
 82.08
 EC Number:
 MDL number:
 NACRES:
 NB.24
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Special Grade
Show More