• USA Home
  • Search Results

  • sulfurtrioxidetriethylaminecomplex1812576101311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sulfur trioxide triethylamine complex

1 Products
Synonym:   NSC 68185, NSC 8168, Triethylamine sulfur trioxide complex
Image of Sulfur trioxide triethylamine complex
CAS Number:
Linear Formula:
 (CH3CH2)3N · SO3
Molecular Weight:
 181.25
 Beilstein/REAXYS Number:
 3993165
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More