• USA Home
  • Search Results

  • sulfurylchloridefluoride118521363784811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sulfuryl chloride fluoride

1 Products
Synonym:   Chlorosulfonyl fluoride, Fluorosulfonyl chloride, Sulfuryl chlorofluoride
Image of Sulfuryl chloride fluoride
CAS Number:
Linear Formula:
 SO2ClF
Molecular Weight:
 118.52
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More