• USA Home
  • Search Results

  • sulindac356413819450211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sulindac

5 Products
Synonym:   (Z)-5-Fluoro-2-methyl-1-[p-(methylsulfinyl)benzylidene]indene-3-acetic acid
Image of Sulindac
CAS Number:
Empirical Formula (Hill Notation):
 C20H17FO3S
Molecular Weight:
 356.41
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More