• USA Home
  • Search Results

  • sunflowerseedoil12345800121611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sunflower seed oil

1 Products
Synonym:   Sunflower seed oil from Helianthus annuus
CAS Number:
 EC Number:
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More