• USA Home
  • Search Results

  • supelcosilicaandc18saturatorcolumns1234598765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Supelco® Silica and C18 Saturator Columns

1 Products
Image of Supelco<SUP>®</SUP> Silica and C18 Saturator Columns
 EC Number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Matrix active group
  Chromatography Mode
  Titertest
  Package quantity
Show More