• USA Home
  • Search Results

  • supelcosilsax1supelguardcartridge1234598765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

SUPELCOSIL SAX1 Supelguard Cartridge

2 Products
 NACRES:
 SB.52
 eCl@ss
 32110501
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Composite Dimension
  Chromatography Mode
  Application(s)
  Particle Dimension
  Package quantity
  Matrix active group
Show More