• USA Home
  • Search Results

  • syringaldehyde1821713496311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Syringaldehyde

3 Products
Synonym:   3,5-Dimethoxy-4-hydroxybenzaldehyde, 4-Hydroxy-3,5-dimethoxybenzaldehyde
Image of Syringaldehyde
CAS Number:
Linear Formula:
 HOC6H2(OCH3)2CHO
Molecular Weight:
 182.17
 Beilstein/REAXYS Number:
 784514
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Agency/Method
  Special Grade
Show More