• USA Home
  • Search Results

  • tadalafil3894017159629511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tadalafil

4 Products
Synonym:   Tadalafil
Image of Tadalafil
CAS Number:
Empirical Formula (Hill Notation):
 C22H19N3O4
Molecular Weight:
 389.40
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
  Manufacturer designation
Show More