• USA Home
  • Search Results

  • tamoxifencitratesalt563645496524111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tamoxifen citrate salt

1 Products
Synonym:   (Z)-1-(p-Dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene
Image of Tamoxifen citrate salt
CAS Number:
Empirical Formula (Hill Notation):
 C26H29NO · C6H8O7
Molecular Weight:
 563.64
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More