• USA Home
  • Search Results

  • tamsulosinhydrochloride4449710646317611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tamsulosin hydrochloride

5 Products
Synonym:   5-[(2R)-2-[[2-(2-Ethoxyphenoxy)ethyl]amino]propyl]-2-methoxybenzenesulfonamide hydrochloride, Tamsulosin hydrochloride
Image of Tamsulosin hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C20H28N2O5S · HCl
Molecular Weight:
 444.97
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
  Application(s)
  Manufacturer designation
Show More