• USA Home
  • Search Results

  • tannicacid170120140155411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tannic acid

5 Products
Synonym:   Gallotannin, Tannin
Image of Tannic acid
CAS Number:
Empirical Formula (Hill Notation):
 C76H52O46
Molecular Weight:
 1701.20
 Beilstein/REAXYS Number:
 8186396
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Loss
  Special Grade
Show More