• USA Home
  • Search Results

  • taqdnapolymerasefromthermusaquaticus12345901290211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Taq DNA Polymerase from Thermus aquaticus

2 Products
Image of Taq DNA Polymerase from <I>Thermus aquaticus</I>
CAS Number:
 Enzyme Commission (EC) Number:
 2.7.7.7  ( BRENDA | IUBMB )
 MDL number:
Popular Documents:  Bulletin (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Color
  Physical Form
  Feature
Show More