• USA Home
  • Search Results

  • tartaricacid150098769411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tartaric acid

2 Products
Synonym:   (2R,3R)-(+)-Tartaric acid, L-(+)-Tartaric acid, L-Threaric acid
Image of Tartaric acid
CAS Number:
Linear Formula:
 HO2CCH(OH)CH(OH)CO2H
Molecular Weight:
 150.09
 Beilstein/REAXYS Number:
 1725147
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
Show More