• USA Home
  • Search Results

  • tartrazine53436193421011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tartrazine

2 Products
Synonym:   Acid Yellow 23
Image of Tartrazine
CAS Number:
Empirical Formula (Hill Notation):
 C16H9N4Na3O9S2
Molecular Weight:
 534.36
 Beilstein/REAXYS Number:
 69850
 Colour Index Number:
 19140
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Special Grade
Show More