• USA Home
  • Search Results

  • taurine1251510735711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Taurine

9 Products
Synonym:   2-Aminoethanesulfonic acid
Image of Taurine
CAS Number:
Linear Formula:
 NH2CH2CH2SO3H
Molecular Weight:
 125.15
 Beilstein/REAXYS Number:
 1751215
 EC Number:
 MDL number:
 NACRES:
 NA.32
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Agency/Method
  pH Value
  UV-Absorption Values
  Manufacturer designation
  Solubility
  Product Line
  Biological Source
  Color
  Assay
  Physical Form
  Special Grade
Show More