• USA Home
  • Search Results

  • taxifolin3042548018211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Taxifolin

2 Products
Synonym:   (2R,3R)-3,3′,4′,5,7-Pentahydroxyflavanone, (2R,3R)-Dihydroquercetin
Image of Taxifolin
CAS Number:
Empirical Formula (Hill Notation):
 C15H12O7
Molecular Weight:
 304.25
 Beilstein/REAXYS Number:
 5299277
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Special Grade
Show More