• USA Home
  • Search Results

  • tazobactamsodiumsalt322278978584211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tazobactam sodium salt

2 Products
Synonym:   CL-298741, YTR-830H, [2S-(2α, 3β, 5α)]-3-Methyl-7-oxo-3(1H,2,3-triazol-1-ylmethyl)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 4,4-dioxide
Image of Tazobactam sodium salt
CAS Number:
Empirical Formula (Hill Notation):
 C10H11N4NaO5S
Molecular Weight:
 322.27
 MDL number:
 NACRES:
 NA.85
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
Show More