• USA Home
  • Search Results

  • tdareagent16220770508011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

TDA Reagent

1 Products
Synonym:   Ferric chloride solution, Iron(III) chloride solution, Tryptophan Deaminase Reagent
Image of TDA Reagent
CAS Number:
Linear Formula:
 FeCl3
Molecular Weight:
 162.20
 MDL number:
 NACRES:
 NA.85
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More