• USA Home
  • Search Results

  • tegafur200171790223711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tegafur

1 Products
Synonym:   5-Fluoro-1-(tetrahydro-2-furyl)uracil, Ftorafur, Tegafur
Image of Tegafur
CAS Number:
Empirical Formula (Hill Notation):
 C8H9FN2O3
Molecular Weight:
 200.17
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More