• USA Home
  • Search Results

  • tellurium127601349480911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tellurium

6 Products
Image of Tellurium
CAS Number:
Empirical Formula (Hill Notation):
 Te
Molecular Weight:
 127.60
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
 eCl@ss:
 38040201
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Particle Dimension
  Physical Form
Show More