• USA Home
  • Search Results

  • telluriumtetrachloride269411002607011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tellurium tetrachloride

1 Products
Image of Tellurium tetrachloride
CAS Number:
Linear Formula:
 TeCl4
Molecular Weight:
 269.41
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More