• USA Home
  • Search Results

  • telmisartan5146214470148411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Telmisartan

4 Products
Synonym:   4′[(1,4′-Dimethyl-2′-propyl[2,6′-bi-1H-benzimidazol]-1′-yl)methyl][1,1′-biphenyl]-2-carboxylic acid, BIBR 277
Image of Telmisartan
CAS Number:
Empirical Formula (Hill Notation):
 C33H30N4O2
Molecular Weight:
 514.62
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Physical Form
  Solubility
Show More