• USA Home
  • Search Results

  • terbium15893744027911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Terbium

17 Products
Image of Terbium
CAS Number:
Empirical Formula (Hill Notation):
 Tb
Molecular Weight:
 158.93
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Weight
  Particle Dimension
  Composite Dimension
  Physical Form
  Manufacturer designation
Show More

Product #
Description
263206 Terbium, chips, 99.9% trace rare earth metals basis,
693715 Terbium, foil, size 25 mm × 25 mm × 1 mm, 99.9% trace rare earth metals basis,
GF21766552 Terbium, powder, 1g, max. particle size 500 micron, 99.9%,
GF68522317 Terbium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%,
GF50413508 Terbium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%,
GF39171517 Terbium, foil, 50x50mm, thickness 0.1mm, as rolled, 99%,
GF16313374 Terbium, foil, 25x25mm, thickness 0.1mm, as rolled, 99%,
GF20819088 Terbium, powder, 2g, max. particle size 500 micron, 99.9%,
GF54878529 Terbium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%,
GF59815794 Terbium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%,
GF77306182 Terbium, rod, 50mm, diameter 6.35mm, cast, 99%,
GF02386166 Terbium, powder, 5g, max. particle size 500 micron, 99.9%,
GF63273731 Terbium, rod, 100mm, diameter 6.35mm, cast, 99%,
GF14307252 Terbium, lump, 25 mm max. lump size, weight 10 g, purity 99%,
GF53377230 Terbium, lump, 25 mm max. lump size, weight 20 g, purity 99%,
GF95900452 Terbium, rod, 2.0 mm diameter, length 50 mm, purity 99%,
GF98270545 Terbium, lump, 25 mm max. lump size, weight 50 g, purity 99%,