• USA Home
  • Search Results

  • terbiumiiitrifluoromethanesulfonate6061314898031811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Terbium(III) trifluoromethanesulfonate

1 Products
Synonym:   Terbium tris(trifluoromethanesulfonate), Terbium tritriflate, Terbium(III) triflate, Trifluoromethanesulfonic acid terbium(III) salt
Image of Terbium(III) trifluoromethanesulfonate
CAS Number:
Linear Formula:
 (CF3SO3)3Tb
Molecular Weight:
 606.13
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More