• USA Home
  • Search Results

  • terpinolene1362358662911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Terpinolene

3 Products
Synonym:   4-Isopropylidene-1-methylcyclohexene, p-Meth-1-en-8-yl-formate, p-Menth-1,4(8)-diene
Image of Terpinolene
CAS Number:
Empirical Formula (Hill Notation):
 C10H16
Molecular Weight:
 136.23
 Beilstein/REAXYS Number:
 1851203
 Council of Europe no.:
 2115
 EC Number:
 FEMA Number:
 3046
 Flavis number:
 1.005
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Agency/Method
  Refractive Index
  Assay
Show More