• USA Home
  • Search Results

  • tertbutylacetate1161654088511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

tert-Butyl acetate

2 Products
Synonym:   TBAc
Image of <I>tert</I>-Butyl acetate
CAS Number:
Linear Formula:
 CH3COOC(CH3)3
Molecular Weight:
 116.16
 Beilstein/REAXYS Number:
 1699506
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More