• USA Home
  • Search Results

  • tertbutyldimethylsilylacetylene140308631861811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(tert-Butyldimethylsilyl)acetylene

1 Products
Synonym:   Ethynyl-tert-butyldimethylsilane
Image of (<I>tert</I>-<WBR>Butyldimethylsilyl)<WBR>acetylene
CAS Number:
Linear Formula:
 (CH3)3CSi(CH3)2C≡CH
Molecular Weight:
 140.30
 Beilstein/REAXYS Number:
 1921466
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More