• USA Home
  • Search Results

  • tertbutylethylether1021763792311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

tert-Butyl ethyl ether

4 Products
Image of <I>tert</I>-Butyl ethyl ether
CAS Number:
Linear Formula:
 C2H5OC(CH3)3
Molecular Weight:
 102.17
 Beilstein/REAXYS Number:
 1731469
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
 eCl@ss:
 39021026
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
  Solubility
  Potency
Show More