• USA Home
  • Search Results

  • tertbutylisocyanate9913160986511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

tert-Butyl isocyanate

1 Products
Synonym:   2-isocyanato-2-methylpropane
Image of <I>tert</I>-Butyl isocyanate
CAS Number:
Linear Formula:
 (CH3)3CN=C=O
Molecular Weight:
 99.13
 Beilstein/REAXYS Number:
 969479
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More