• USA Home
  • Search Results

  • tertbutylmethylether8815163404411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

tert-Butyl methyl ether

19 Products
Synonym:   MTBE, Methyl tert-butyl ether
Image of <I>tert</I>-Butyl methyl ether
CAS Number:
Linear Formula:
 (CH3)3COCH3
Molecular Weight:
 88.15
 Beilstein/REAXYS Number:
 1730942
 EC Number:
 MDL number:
 NACRES:
 NA.07
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Purity Grade
  Package quantity
  Shelf Life
  Solubility
  Color
  Refractive Index
  UV-Absorption Condition
  Viscosity
  Transition temperature
  Explosion limit
  Autoignition temp
  Density
  Transmittance
  Potency
  Melting Point
  Assay
  Vapor pressure
  UV-Absorption Values
Show More

Product #
Description
306975 tert-Butyl methyl ether, anhydrous, 99.8%,
34875 tert-Butyl methyl ether, for HPLC, ≥99.8%,
650560 tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%,
443808 tert-Butyl methyl ether, ACS reagent, ≥99.0%,
179787 tert-Butyl methyl ether, reagent grade, ≥98%,
675407 tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%,
20256 tert-Butyl methyl ether, puriss. p.a., ≥99.5% (GC),
293210 tert-Butyl methyl ether, HPLC grade, for HPLC, 99.8%,
320196 tert-Butyl methyl ether, reagent grade, 98%,
08603 tert-Butyl methyl ether, analytical standard,
1.01845 tert-Butyl methyl ether, for liquid chromatography LiChrosolv®,
1.01849 tert-Butyl methyl ether, for analysis EMSURE® ACS,
1.01843 tert-Butyl methyl ether, EMPLURA®,
1.01995 tert-Butyl methyl ether, for gas chromatography ECD and FID SupraSolv®,
1.01984 tert-Butyl methyl ether, for spectroscopy Uvasol®,
1083303 tert-Butyl methyl ether, United States Pharmacopeia (USP) Reference Standard,
8.44008 tert-Butyl methyl ether, for synthesis,
PHR1560 tert-Butyl Methyl Ether, Pharmaceutical Secondary Standard; Certified Reference Material,
RTC000083 tert-Butyl Methyl Ether, Pharmaceutical Secondary Standard; Certified Reference Material,