• USA Home
  • Search Results

  • testosterone288425822011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Testosterone

6 Products
Synonym:   17β-Hydroxy-3-oxo-4-androstene, 17β-Hydroxy-4-androsten-3-one, 4-Androsten-17β-ol-3-one, trans-Testosterone
Image of Testosterone
CAS Number:
Empirical Formula (Hill Notation):
 C19H28O2
Molecular Weight:
 288.42
 Beilstein/REAXYS Number:
 1915399
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Assay
  Anal. Standard Form
  Physical Form
  Application(s)
  Manufacturer designation
  Special Grade
Show More