• USA Home
  • Search Results

  • testosteroneacetate33046104569811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Testosterone acetate

1 Products
CAS Number:
Empirical Formula (Hill Notation):
 C21H30O3
Molecular Weight:
 330.46
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
NMIS042 Testosterone acetate, New