• USA Home
  • Search Results

  • tetraammineplatinumiinitrate387212063412211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tetraammineplatinum(II) nitrate

2 Products
Image of Tetraammineplatinum(II) nitrate
CAS Number:
Linear Formula:
 [Pt(NH3)4](NO3)2
Molecular Weight:
 387.21
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Reaction Suitability
Show More