• USA Home
  • Search Results

  • tetrabutylammoniumbenzoate363581881989111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tetrabutylammonium benzoate

1 Products
Image of Tetrabutylammonium benzoate
CAS Number:
Linear Formula:
 (CH3CH2CH2CH2)4N(OCOC6H5)
Molecular Weight:
 363.58
 Beilstein/REAXYS Number:
 4070140
 MDL number:
 NACRES:
 NB.61
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More