• USA Home
  • Search Results

  • tetrabutylammoniumdifluorotriphenylstannate6304813935388111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tetrabutylammonium difluorotriphenylstannate

1 Products
Synonym:   Tetrabutylammonium difluorotriphenyltin
Image of Tetrabutylammonium difluorotriphenylstannate
CAS Number:
Linear Formula:
 (CH3CH2CH2CH2)4N[(C6H5)3SnF2]
Molecular Weight:
 630.48
 Beilstein/REAXYS Number:
 8668854
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More