• USA Home
  • Search Results

  • tetrabutylammoniumiodide3693731128411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tetrabutylammonium iodide

3 Products
Synonym:   TBAI
Image of Tetrabutylammonium iodide
CAS Number:
Linear Formula:
 (CH3CH2CH2CH2)4N(I)
Molecular Weight:
 369.37
 Beilstein/REAXYS Number:
 3916152
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Melting Point
  UV-Absorption Values
Show More