• USA Home
  • Search Results

  • tetracaine264369424611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tetracaine

4 Products
Synonym:   4-(Butylamino)benzoic acid 2-(dimethylamino)ethyl ester
Image of Tetracaine
CAS Number:
Empirical Formula (Hill Notation):
 C15H24N2O2
Molecular Weight:
 264.36
 EC Number:
 MDL number:
 NACRES:
 NA.32
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
Show More