• USA Home
  • Search Results

  • tetrachloroethylene1658312718411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tetrachloroethylene

8 Products
Synonym:   PCE, Perchloroethylene, Tetrachloroethylene
Image of Tetrachloroethylene
CAS Number:
Linear Formula:
 CCl2=CCl2
Molecular Weight:
 165.83
 Beilstein/REAXYS Number:
 1361721
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Vapor pressure
  Potency
  Melting Point
  Density
  Color
  Solubility
  UV-Absorption Values
  Special Grade
  Physical Form
  Application(s)
Show More