• USA Home
  • Search Results

  • tetracycline44443anhydrousbasis6054811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tetracycline

2 Products
Image of Tetracycline
CAS Number:
Empirical Formula (Hill Notation):
 C22H24N2O8 · xH2O
Molecular Weight:
 444.43 (anhydrous basis)
 Beilstein/REAXYS Number:
 2230417
 EC Number:
 MDL number:
 NACRES:
 NA.85
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More