• USA Home
  • Search Results

  • tetracyclinehydrochloride480906475511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tetracycline hydrochloride

8 Products
Image of Tetracycline hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C22H24N2O8 · HCl
Molecular Weight:
 480.90
 Beilstein Registry Number:
 3844873
 EC Number:
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
  Titertest
  Product Line
  Anal. Standard Form
  Special Grade
  Color
  Biological Source
  Mode of Action
  Physical Form
Show More