• USA Home
  • Search Results

  • tetradecafluorohexane3380435542011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tetradecafluorohexane

2 Products
Synonym:   Perfluorohexane
Image of Tetradecafluorohexane
CAS Number:
Linear Formula:
 CF3(CF2)4CF3
Molecular Weight:
 338.04
 Beilstein/REAXYS Number:
 1802113
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More