• USA Home
  • Search Results

  • tetradecanophenone28847449705611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tetradecanophenone

1 Products
Image of Tetradecanophenone
CAS Number:
Linear Formula:
 C6H5CO(CH2)12CH3
Molecular Weight:
 288.47
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
298611 Tetradecanophenone, 95%,