• USA Home
  • Search Results

  • tetradecylphosphonicacid27837467175411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tetradecylphosphonic acid

1 Products
Synonym:   TDPA, TPA
Image of Tetradecylphosphonic acid
CAS Number:
Linear Formula:
 CH3(CH2)13P(O)(OH)2
Molecular Weight:
 278.37
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More