• USA Home
  • Search Results

  • tetraethylammoniumchloride165705634811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tetraethylammonium chloride

3 Products
Synonym:   TEA chloride
Image of Tetraethylammonium chloride
CAS Number:
Linear Formula:
 (C2H5)4N(Cl)
Molecular Weight:
 165.70
 Beilstein/REAXYS Number:
 3563247
 EC Number:
 MDL number:
 NACRES:
 NA.32
 PubChem Substance ID:
Popular Documents:  Product Information Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  UV-Absorption Values
  Application(s)
  Solubility
  Special Grade
Show More