• USA Home
  • Search Results

  • tetraethylammoniumptoluenesulfonate3014473344811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tetraethylammonium p-toluenesulfonate

1 Products
Synonym:   p-Toluenesulfonic acid tetraethylammonium salt
Image of Tetraethylammonium <I>p</I>-toluenesulfonate
CAS Number:
Linear Formula:
 (C2H5)4N(CH3C6H4SO3)
Molecular Weight:
 301.44
 Beilstein/REAXYS Number:
 4303354
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More